missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wortmannin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 mg
5 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 234.0°C to 240.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 1 mg |
Beilstein | 19, IV, 3134 |
Merck Index | 15, 10251 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in methanol (5mg/mL),ethanol,,chlorform,dmso (25mg/mL) |
Formula Weight | 428.4 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Wortmannin
- Danger
- Acute toxicity Category 1
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'