missing translation for 'onlineSavingsMsg'
Learn More

Wood’s Metal, Pure, M.P.70°C

Catalog Number 10142210
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
76093-98-6
Bi·Cd·Pb·Sn
198175
This item is not returnable. View return policy
76093-98-6
Bi·Cd·Pb·Sn
198175
Color Gray
Melting Point 70°C
Quantity 100 g
Packaging Plastic powder jar
Form Solid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Wood's metal m.p. ca. 70°C
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1A
  • Reproductive toxicity on or via lactation
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H362-May cause harm to breast-fed children.
  • H372-Causes damage to organs.
  • H360FD-May damage fertility. May damage the unborn child.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P263-Avoid contact during pregnancy/while nursing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'