missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Wnt-2b Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Wnt-2b |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
Wnt-2b Polyclonal specifically detects Wnt-2b in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Wnt-2b | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human, Mouse | |
Protein Wnt-13, protein Wnt-2b, wingless-type MMTV integration site family, member 2B, Wnt-13, WNT13XWNT2, Xenopus, homolog of, Wnt2b, XWNT2, XWNT2wingless-type MMTV integration site family, member 13 | |
WNT2B | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
O70283 | |
7482 | |
E. coli-derived recombinant mouse Wnt-2b Leu64-Glu104 and Arg251-Ala312 Accession # O70283 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects recombinant human and recombinant mouse Wnt-2b in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Wnt-2 is observed and less than 5% cross-reactivity with rhWnt-7a, -9a, recombinant mouse Wnt-1, -3a, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, and -16 is observed. |
For Research Use Only