missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Wnt-10b Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Wnt-10b |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Wnt-10b Polyclonal specifically detects Wnt-10b in Mouse samples. It is validated for Western Blot.Specifications
Wnt-10b | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, Wnt-12, WNT12 | |
WNT10B | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P48614 | |
7480 | |
E. coli-derived recombinant mouse Wnt-10b peptide Leu42-Glu82, Arg263-Arg332 Accession # P48614 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Wnt-10b in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Wnt-3a, rmWnt-4, rmWnt-5a, and rmWnt-8a is observed. |
For Research Use Only