missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ WISP1 (Human) ELISA Kit

For the Quantitative detection of Human WISP1.

Brand:  Abnova™ KA4255.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16098655

  • £527.81 / 1 set


Estimated Shipment
22-06-2020
Add to basket

Description

Description

  • WISP1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human WISP1.
Specifications

Specifications

Human
8840
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
WISP1
WNT1 inducible signaling pathway protein 1
Kit
Human
1 Kit
Videos
Documents