Learn More
AZOWIPE™ Wipes
Bactericidal hard surface disinfectant wipes. AZOWIPE™ Wipes are convenient for use in all clinical environments and are an important part of a robust infection control protocol.
Brand: Vernacare 81123
Code : 03
Additional Details : Weight : 9.50000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Made from a viscose rayon nonwoven material strong and durable in use
- Foil sealed - prevents evaporation in storage
- Suitable for all hard surfaces - one product from a single source
- Contains NO preservative - no smearing or streaking when in use
- Specially designed canister - easy to dispense wipes
- 70% Solution of Isopropyl Alcohol BP
- AZOWIPE - for the disinfection of hard surfaces
- Dimensions: 200mm x 220mm
Specifications
Wipes | |
220 x 200 mm |