missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Winter Proof Driver Gloves
Click to view available options
Size:
8
9
10
Description
- Water repellent cowhide grain leather glove
- Brushed fleece lining
- Elasticated at the back
- The leather has been enriched by a special tanning process to give the glove superior water repellent properties
- The lining increases the glove's comfort and cold insulation
- Elasticated at the wrist, the glove offers added support
- The winged thumb makes the glove more anatomical for greater ease of use
- The water repellent treatment enables the glove to be used in damp environments
- Cowhide leather provides good mechanical resistance
- Assembly ensures a longer life while reducing the number of seams
Specifications
Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388:2016 Mechanical hazards, EN 511-2006 Cold protection |
Coating Material | Cowhide Leather |
Color | Black |
Size | 9 |
Thickness (Metric) | 0.8 to 1.1 mm |
Length (Metric) | 26.5 cm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'