missing translation for 'onlineSavingsMsg'
Learn More

WIN 64338 hydrochloride, Tocris Bioscience™

Bradykinin B2 antagonist

Brand:  Tocris (Bio-Techne) 1057/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458403

  • £175.56 / 10mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

The first potent, non-peptide, competitive bradykinin B2 receptor antagonist. In organ bath studies, WIN 64338 inhibits [3H]-bradykinin binding on guinea pig trachea with nanomolar affinity but is not active in the rabbit aorta (the classical bradykinin B1 preparation).
Specifications

Chemical Identifiers

163727-74-0
783.948
win 64338 hydrochloride, s-4-2-bis cyclohexylamino methyleneamino-3-2-naphthalenyl-1-oxopropylamino benzyl tributyl phosphonium chloride hydrochloride, phosphonium,tributyl 4-2s-2-cyclohexylamino cyclohexylimino methyl amino-3-2-naphthalenyl-1-oxopropyl amino phenyl methyl-,chloride 1:1
tributyl-[[4-[[(2S)-2-[(N,N'-dicyclohexylcarbamimidoyl)amino]-3-naphthalen-2-ylpropanoyl]amino]phenyl]methyl]phosphanium;chloride;hydrochloride
C45H69Cl2N4OP
YYJGBEZPVOUBMJ-KRFCICRISA-N
9832172
CCCC[P+](CCCC)(CCCC)CC1=CC=C(C=C1)NC(=O)C(CC2=CC3=CC=CC=C3C=C2)NC(=NC4CCCCC4)NC5CCCCC5.Cl.[Cl-]

Specifications

WIN 64338 hydrochloride
163727-74-0
win 64338 hydrochloride, s-4-2-bis cyclohexylamino methyleneamino-3-2-naphthalenyl-1-oxopropylamino benzyl tributyl phosphonium chloride hydrochloride, phosphonium,tributyl 4-2s-2-cyclohexylamino cyclohexylimino methyl amino-3-2-naphthalenyl-1-oxopropyl amino phenyl methyl-,chloride 1:1
CCCC[P+](CCCC)(CCCC)CC1=CC=C(C=C1)NC(=O)C(CC2=CC3=CC=CC=C3C=C2)NC(=NC4CCCCC4)NC5CCCCC5.Cl.[Cl-]
783.948
783.95
10mg
C45H69Cl2N4OP
YYJGBEZPVOUBMJ-KRFCICRISA-N
tributyl-[[4-[[(2S)-2-[(N,N'-dicyclohexylcarbamimidoyl)amino]-3-naphthalen-2-ylpropanoyl]amino]phenyl]methyl]phosphanium;chloride;hydrochloride
9832172
>98%
Videos