missing translation for 'onlineSavingsMsg'
Learn More

Wilmad-LabGlass™ SP Scienceware™ Rotor and Caps for Bruker MAS Probe

Provide solution for analysis of solid samples

£17.54 - £61.70

Specifications

For Use With (Application) Solid state NMR spectrometers
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Model Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Model Price Quantity & Availability  
15399028
View Documents Promotion Details
Wilmad-LabGlass™
66809-0581
For Bruker MAS 3.2mm Probe 520-1A
£61.70
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
 
15369028
View Documents Promotion Details
Wilmad-LabGlass™
66800-0545
For Bruker MAS 7mm Probe 528-PP-4VAR This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Manufactured from high-purity Zirconia
  • Caps fitted with O-rings for improved sealing
  • MAS Probe and caps compatible for most solid state NMR spectrometers
  • Thoroughly inspected before and after precision-machining process ensures no material irregularities
  • Spin testing performed to highest specified spinning speed, assuring performance without overspinning the rotor
     
Specifications

Specifications

Solid state NMR spectrometers
Videos
SDS
Documents

Documents

Product Certifications