missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Pyrex™ Capillary Tubes

Manufactured from Pyrex™ and sealed at one end. Bel-Art SP Scienceware™ Wilmad™ Pyrex™ Capillary Tubes provide a low cost solution for small volume NMR measurement.

Brand:  Bel-Art™ 66000-0071

Code : 03

Additional Details : Weight : 0.04310kg

 View more versions of this product

Product Code. 15348648

  • £37.75 / Pack of 10


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Made of ASTM Type 1 Class A glass
  • Tested at 400 MHz field
Specifications

Specifications

Capillary tube
127mm
1.3mm
10Pack
Pyrex™ glass
1.7mm
400MHz
WG-1364-1.7-5
Videos