missing translation for 'onlineSavingsMsg'
Learn More

Bel Art Products 5PK NMR Sample Tube 9 inch, 7.5mm thin wall .

Brand:  Bel Art Products 66350-7595

5PK NMR Sample Tube 9 inch, 7.5mm thin wall .

Code : 03

Additional Details : Weight : 0.04310kg

Product Code. 15388888

  • £160.33 / Pack of 5


Call for latest availability
+44 (0)1509 555 500
Add to basket