missing translation for 'onlineSavingsMsg'
Learn More

Wilmad™ SP Scienceware™ 15mm O.D. Thin Walled Precision NMR Tube

To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts

Brand:  Wilmad™ 66300-1585

Code : 03

Additional Details : Weight : 0.04310kg

 View more versions of this product

Product Code. 15308838

  • POA / Pack of 5

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.

Alert:

Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.


Specifications

Specifications

203.2mm
13.470±0.013mm
For experiments requiring critical shimming quality (high/ultrahigh field, non-spinning, multi-dimensional, multi-nuclei, DNP experiments and studies involving biological samples)
515-3PP-8-5
15.065±0.008mm
300MHz
0.76mm
Videos