missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Wif1 (Mouse) ELISA Kit

For the Quantitative detection of Mouse Wif1.

Brand:  Abnova™ KA5157.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16079395

  • £527.81 / 1 set


Estimated Shipment
22-06-2020
Add to basket

Description

Description

  • Wif1 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of mouse Wif1 in cell culture supernates, cell lysates, serum and plasma (heparin or EDTA).
Specifications

Specifications

Mouse
24117
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
Wif1
Wnt inhibitory factor 1
Kit
Mouse
1 Kit
Videos
Documents