missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ WIF1 (Human) ELISA Kit

For the Quantitative detection of Human WIF1.

Brand:  Abnova™ KA5158.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16089395

  • £508.00 / 1 set


Estimated Shipment
17-06-2020
Add to basket

Description

Description

  • WIF1 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative measurement of Human WIF1 in cell culture supernates, cell lysates, serum and plasma (heparin or EDTA).
Specifications

Specifications

Human
11197
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
WIF1
WNT inhibitory factor 1
Kit
Human
1 Kit
Videos
Documents