missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Wide Neck Soda Lime Glass Reagent Bottless with Stopper
Manufactured from soda lime glass. DURAN™ Wide Neck Soda Lime Glass Reagent Bottless with Stopper is DIN 12039 certified.
Brand: Duran™ 1039353
Code : 01
Additional Details : Weight : 0.29704kg
Description
- Wide neck
- Clear
- With individually ground glass stopper
- Soda-lime-glass
- DIN 12039
Specifications
250mL | |
10Pack | |
Bottle, Reagent | |
129mm |
Soda Lime Glass | |
NS 34/24 | |
71mm |