missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck Soda Lime Glass Reagent Bottless with Stopper

Manufactured from soda lime glass. DURAN™ Wide Neck Soda Lime Glass Reagent Bottless with Stopper is DIN 12039 certified.

Brand:  Duran™ 1039354

Code : 01

Additional Details : Weight : 0.47000kg

 View more versions of this product

Product Code. 12850954

  • £33.75 / Each


In Stock 22
Add to basket

Description

Description

  • Wide neck
  • Clear
  • With individually ground glass stopper
  • Soda-lime-glass
  • DIN 12039
Specifications

Specifications

500mL
10Pack
Bottle, Reagent
164mm
Soda Lime Glass
NS 45/40
86mm
Videos
Documentation