missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck PU Coated Glass Laboratory Bottle with Screw Cap and Pouring Ring

Brand:  Duran™ 1160152

Code : A0

Additional Details : Weight : 4.80000kg

Product Code. 11796988

  • POA / Pack of 10

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

500mL
Polypropylene
PU Coating
Storage, transport and safe handling of hazardous substances
Glass
10
10Pack
Bottle
Blue
101mm
Wide
USP Standard, EU Design 381983 Certified
Yes
Clear
Screw Cap and Pouring Ring
140°C
Bottle
Yes
Yes
101mm
148mm
Quick Release Closure
Videos