missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck Glass Reagent Bottle with Flat Head Stopper

Brand:  Duran™ 211651402

Code : A0

Additional Details : Weight : 0.34000kg

 View more versions of this product

Product Code. 11756928

  • £300.00 / Pack of 10


Estimated Shipment
01-02-2018
Add to basket

Specifications

Specifications

25mL
Borosilicate Glass
Bottle, Reagent
36mm
Narrow
ISO 4796-2
10Pack
Yes
64mm
Videos