missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck Glass Reagent Bottle with Flat Head Stopper

Brand:  Duran™ 211855409

Code : A0

Additional Details : Weight : 7.75000kg

 View more versions of this product

Product Code. 11796253

  • £725.00 / Pack of 10


Estimated Shipment
06-06-2018
Add to basket

Specifications

Specifications

1000mL
Clear
Standard Ground Joint, Glass Flat-Head Stopper
60/46
Bottle, Reagent
201mm
ISO 4796-2, USP Standard
Suitable for storage of aggressive media, which could attack plastic parts.
Glass
10Pack
107mm
Wide Mouth
Videos