missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck Glass Reagent Bottle with Flat Head Stopper

Brand:  Duran™ 211852403

Code : A0

Additional Details : Weight : 1.40000kg

 View more versions of this product

Product Code. 11776253

  • £324.00 / Pack of 10


Estimated Shipment
15-06-2018
Add to basket

Specifications

Specifications

100mL
Clear
Standard Ground Joint, Glass Flat-Head Stopper
29/22
Bottle, Reagent
97mm
ISO 4796-2, USP Standard
Suitable for storage of aggressive media, which could attack plastic parts.
Glass
10Pack
52mm
Wide
Videos