missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck Flask

Brand:  Duran™ 217416803

Code : A0

Additional Details : Weight : 0.82000kg

 View more versions of this product

Product Code. 10709491

  • £63.50 / Each


Estimated Shipment
30-01-2018
Add to basket

Specifications

Specifications

Round
185mm
Ideal for heating applications
Wide
DIN ISO 24450, USP Standard
3000mL
260mm
Videos