missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Wide Neck Borosilicate Glass Reagent Bottle

Brand:  Duran™ 211845408

Code : A0

Additional Details : Weight : 5.40000kg

 View more versions of this product

Product Code. 11736263

  • £555.00 / Pack of 10


Estimated Shipment
25-06-2018
Add to basket

Specifications

Specifications

1000mL
Standard Ground Joint
500°C
10Pack
Yes
104mm
ISO 4796-2, USP Standard
Borosilicate Glass
10Pack
Bottle, Reagent
Yes
201mm
Videos