Thermo Scientific™ Whole Cell Stains

Provide excellent staining for high-content screening assays and other fluorescent cellular imaging methods involving fluorescence microscopy.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 8403402

UNSPSC: 41106513

Code: PN

Additional Details:
Additional Details: Weight: 0.80000kg Transport: UN number : 3261 Chem class : 1a Pack group : III



Product Code. 10096853

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Description Cellomics Whole Cell Stain Red
Quantity 5 × 96 tests
Target Whole cell (red); Ex/Em = 654/673nm

These stains are intense, highly photostable and match the output wavelengths of common fluorescence instrumentation. Effective image analysis in cell-based assays and fluorescence microscopy of the localization of specific targets requires fluorescent labeling of the entire cells, nuclei or major cellular components. Whole cell stains identify cell numbers, cell borders and cell morphologies as useful primary objects for assessment of specific protein targets and their redistribution in response to treatment conditions.
  • Provide excellent staining for HCA and fluorescence microscopy
  • Intense, highly photostable and match the output wavelengths of common fluorescence instrumentation
  • When the whole cell is the primary object used to identify and count individual cells, effectively distinguish intact cells from bordering cells (with analysis by appropriate software)

Includes:

Fluorescent stains to detect whole cells (blue, green, orange or red)

Requires:

Fluorescence microscope, Thermo Scientific™ Cellomics™ ArrayScan™ VTI or other HCS reader

Documentation

Technical documents