Learn More
3M™ White Polyethylene Laminate Category I Protective Coveralls
Specifications
Product Type | Protective Coveralls |
Waist Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 2-panel |
Protection Class | Category I |
Height (Metric) | 179 to 187 cm |