missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ White Plastic General Purpose Trays
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.81000kg
Description
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifications
570 x 340mm | |
570 mm | |
Tray | |
Safe containment of spillages, lab bench protection | |
1 Each |
White | |
Plastic | |
340 mm | |
Liner | |
Rectangular |