missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Screw Caps for Glass Bottles and Vials

Catalog Number p-4608864
Click to view available options
Neck Finish:
15-425
18-400
24-400
For Use With (Equipment):
Glass EPA vial
Glass bottle or vial
No. per Case:
144 Cs.
200 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

With clear surface for writing

  • Available with different liners (PTFE/silicone, foamed polyethylene, poly-vinyl, PTFE faced foamed polyethylene)
  • With choice of open or closed top

Caps with PTFE/silicone liner:

  • Provides heat resistance
  • Glass filled
  • Suited for EPA samples
Caps with foamed polyethylene liner:
  • Chemical resistant
  • Tight seal
Caps with poly-vinyl liner:
  • With moisture barrier
  • Suited for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Caps with PTFE faced foamed polyethylene liner:
  • Chemical resistant
  • Provides compressibility and sealing properties
  • Suited for high purity chemicals, strong acids and solvents

Closure Liner PTFE/Silicone
For Use With (Equipment) Glass EPA vial
Color White
Material Polypropylene
No. per Case 200 Cs.
Neck Finish 24-400
Certifications/Compliance ISO 9001
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'