missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials

Catalog Number 10262983
Click to view available options
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
No. per Pack:
100/Cs.
1000/Cs.
250/Cs.
500/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

PTFE faced silicone liner ideal for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

  • Autoclavable

Closure Liner PTFE/Silicone
For Use With (Equipment) 2 and 4mL Vial
Color White
Material Polypropylene
No. per Case 1000 Cs.
No. per Pack 1000/Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'