missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Material:
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Description
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
- PTFE faced foamed polyethylene liner offers chemical resistance
- Polyethylene foam provides compressibility and sealing properties
- Good for high purity chemicals, strong acids, and solvents
- For usewith glass bottles or vials
- Polypropylene
- Screw cap
- White
Specifications
Closure Liner | PTFE/Polyethylene |
For Use With (Equipment) | Glass bottle or vial |
Color | White |
Material | Polypropylene |
No. per Case | 72 Cs. |
Neck Finish | 43-400 |
Certifications/Compliance | ISO 9001 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'