missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners

Catalog Number 11798829
Click to view available options
Neck Finish:
22-400
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
89-400
No. per Case:
144 Cs.
48 Cs.
72 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products

  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
Closure Liner Polyvinyl
For Use With (Equipment) Round bottle
Product Type Polypropylene Cap
Color White
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 58-400
Certifications/Compliance ISO 9001
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'