missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Wheaton™ Tenbroeck Tissue Grinder

With expanded reservoir and pouring lip

Brand:  Wheaton™ 357428

Code : 03

Additional Details : Weight : 0.72570kg

 View more versions of this product

Product Code. 10698643

  • £199.06 / Pack of 2


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

40mL
Tenbroeck tissue grinder
140mm
1.35 oz., 40mL
Borosilicate Glass
320mm
32mm
Videos
Documentation

Technical Documents