missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper
Provide airtight covers
Brand: DWK Life Sciences 224100-060
Code : A0
Additional Details : Weight : 1.00000kg
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Red | |
1.3 cm | |
5 mm | |
Rubber | |
40 Durometer |
Sleeve Stopper | |
Multi-puncture applications | |
11 mm | |
Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials | |
Yes |