Learn More
DWK Life Sciences Wheaton™ BioStir™ Magnetic Stirrers
Ideal for general mixing and difficult-to-dissolve solutes. Wheaton™ BioStir™ Magnetic Stirrers are available in single or 4-place units.
POA
Description
- Programmable
- 5 to 200 RPM
- Remote control and alarm capable
- Waterproof stainless steel housing
- Operating temperature range of 15°C to 40°C
Specifications
Slow | |
Micro-stir |