missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers
Click to view available options
Neck Finish:
15-425
20-400
22-400
24-400
28-400
33-400
Description
- Poly-Seal liner is manufactured from low density polyethylene
- Phenolic
- Screw cap
- Color: Black
Specifications
Closure Liner | Polyseal |
For Use With (Equipment) | Redi-pak™ container |
Color | Black |
Material | Phenolic Resin |
No. per Case | 144 Cs. |
Neck Finish | 33-400 |
Certifications/Compliance | ISO 9001 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'