Learn More
DWK Life Sciences Wheaton™ Potter-Elvehjem Tissue Grinders With Radial Serrations
Serrated PTFE pestle disperses homogenate into the mortar cylinder more efficiently
Brand: DWK Life Sciences 357974
Additional Details : Weight : 0.92990kg
Description
Grinders are precisely constructed with completely interchangeable parts, allowing for easier replacement of parts.
- Mortar made from low extractable borosilicate glass
- Radial serrated PTFE pestle
- Interchangeable parts
- Autoclavable
Order Info
Grinding chamber clearance is 0.004 in. to 0.006 in. (0.1 to 0.15mm). Depth of the serrations is 0.020 in. to .025 in. . Diameter of the stainless steel shaft is 0.25 in. (6.3mm).
Specifications
0.51 in. | |
0.16 oz. | |
Borosilicate Glass, PTFE | |
8.62 in. | |
2.59 in. | |
0.16 oz., 5mL |
13mm | |
5 mL | |
Tissue Grinder | |
219mm | |
66mm |