missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles

Catalog Number p-76237
Click to view available options
Color:
Black
White
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with Wheaton Glass Diagnostic Bottles

  • Unlined
  • Open- and closed-top caps
  • 20-400 screw thread
  • Autoclavable

Closure Type Open Top Cap
For Use With (Equipment) Screw neck diagnostic bottle
Color Black
Material Polypropylene
No. per Case 300 Cs.
Neck Finish 20-400
Certifications/Compliance ISO 9001
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'