missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Open Top Screw Caps with Silicone Liner

Catalog Number p-4450881
Click to view available options
Neck Finish:
13-425
15-425
Color:
Black
White
No. per Case:
1000 Cs.
250 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Open top screw caps have a bonded PTFE faced silicone liner that provides access with a syringe.

  • Can withstand autoclaving / sterilization
Closure Liner PTFE/Silicone
For Use With (Equipment) Glass bottle or vial
Product Type Open Top Screw Cap
Color Black
Material Phenolic Resin
No. per Case 250 Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'