Learn More
DWK Life Sciences Wheaton™ Open Top Screw Caps with Silicone Liner
Brand: DWK Life Sciences W240851
Additional Details : Weight : 0.10000kg
Description
- Can withstand autoclaving / sterilization
Specifications
ISO 9001 | |
Glass bottle or vial | |
Phenolic | |
Open Top Screw Cap |
PTFE/Silicone | |
Black | |
1000 Cs. | |
15-425 |