missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Media/Laboratory Bottle Caps
Click to view available options
Neck Finish:
33-430
38-430
Material:
Polyethylene
Rubber
Description
- Black phenolic caps with screw thread
- Low Density Polyethylene (Not autoclavable), Rubber lined (Autoclavable)
Specifications
Closure Liner | 14B Rubber |
For Use With (Equipment) | Media bottle |
Color | Black |
Material | Rubber |
No. per Case | 200 Cs. |
Neck Finish | 38-430 |
Certifications/Compliance | ISO 9001 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'