missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Media/Laboratory Bottle Caps

Catalog Number 11758879
Click to view available options
Neck Finish:
33-430
38-430
Material:
Polyethylene
Rubber
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with Wheaton media bottles

  • Black phenolic caps with screw thread
  • Low Density Polyethylene (Not autoclavable), Rubber lined (Autoclavable)

Closure Liner 14B Rubber
For Use With (Equipment) Media bottle
Color Black
Material Rubber
No. per Case 200 Cs.
Neck Finish 33-430
Certifications/Compliance ISO 9001
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'