missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Magna-Flex™ Spinner Flasks

Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension

Brand:  Wheaton™ 356832

Code : 50

Additional Details : Weight : 1.17930kg

 View more versions of this product

Product Code. 10488512

  • £257.97 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Borosilicate glass flasks
  • Two large side arms with screw-cap closures
  • Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
  • Graduated
  • Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
  • Autoclavable flasks and impellers for selected sizes can also be ordered separately
  • U.S. Patent No. 3,854,704

Specifications

Specifications

Yes
4.5cm
Borosilicate Glass
Flask with impeller
10 to 44cm
500mL
Yes
Videos
Documentation View accessory products