missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Magna-Flex™ Spinner Flasks

Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension

Brand:  Wheaton™ 356837

Code : 50

Additional Details : Weight : 4.08230kg

 View more versions of this product

Product Code. 10789341

  • £480.29 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Yes
4.5cm
Borosilicate Glass
Flask with impeller
10 to 44cm
3000mL
Yes
Videos
Documentation