missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (13mm)

Catalog Number 12496031
Click to view available options
Color:
Blue
Gray
Green
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

13mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000
For Use With (Equipment) Plastic dropping bottle
Product Type Dropper Tips
Color Red
Material LDPE
No. per Case 1000 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'