Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
Brand: DWK Life Sciences 356899
Code : A0
Additional Details : Weight : 0.45360kg
Description
- This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
250mL Celstir™ Spinner Flask | |
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle | |
Impeller |