missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ HPLC Mobile-Phase Reservoirs
Suitable for use as mobile phase and filtration reservoirs, as well as general sample and buffer storage
£200.00 - £459.00
Specifications
Certifications/Compliance | USP Type I and ASTM E 438 Type I, Class A |
---|---|
Material | Glass |
Type | HPLC Mobile-phase Reservoir |
For Use With (Application) | Ideal for mobile phase or filtration reservoir |
Bottom Shape | Flat |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
12614813
|
DWK Life Sciences
264710 |
2000 mL | 138 mm | 246 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12624813
|
DWK Life Sciences
264711 |
5000 mL | 186 mm | 318 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Manufactured from Wheaton 33™ low extractable borosilicate glass, which meets ASTM Type I, Class A and USP Type I standards
- Plastic safety coating with an integral UV-blocking agent helps retain glass and liquid in case of breakage and block virtually all UV light up to 385nm
- Screw thread: GL 45
- Autoclavable up to 121°C at 15-20 psig for 15-20 minutes
- Flat bottom
Specifications
USP Type I and ASTM E 438 Type I, Class A | |
HPLC Mobile-phase Reservoir | |
Flat |
Glass | |
Ideal for mobile phase or filtration reservoir |