Learn More
Wheaton™ E-Z Ex-Traction Type I Glass MicroTubes
Brand:  Wheaton™ W224656
Code : 04
Additional Details : Weight : 1.36080kg
 View more versions of this product
Product Code. 11758979
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Yes
Clear
Borosilicate Glass
Locking lid
1.4mL
7.6mm
Ideal for automated storage and retrieval equipment and multichannel pipettes
Flat
Videos