Learn More
DWK Life Sciences Wheaton™ Crimpenstein™ Electric Crimper/Decapper - Non-Standard Voltage
Lightweight and easy to operate
Brand:  DWK Life Sciences W225644-C
Code : NEW
Additional Details : Weight : 2.72160kg
 View more versions of this product
Product Code. 15288474
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Each Crimpenstein comes with a detachable power cord, a convenient carrying/storage case, a crimper and deapper jaw set or an individual crimper or decapper jaw.
Description
Description
Wheaton Crimpenstein Electric Crimper/Decapper
  • Easily attaches and removes aluminum seals on crimp top/serum finish bottles and vials to improve productivity
  • Adjustable jaw intensity with memory allows user to regulate crimper and decapper jaw action
  • Quick-change jaw-sets available in 11, 13 and 20mm sizes allow for crimping and decapping of different size bottles and vials
  • Lightweight and easy to operate enables semiautomatic operation for extended periods with less effort
  • Electric powered for quick setup and high volume operation
For information on using the Crimpenstein with flip cap aluminum seals, visit www.wheatonsci.com/crimp. For information on Wheaton crimp top/serum finish bottles and vials, aluminum seals and rubber stoppers, visit www.fishersci.com.
Optional sturdy stand (Part No. W225630) holds the Crimpenstein in a fixed vertical position for operations where vials need to be handled one at a time.
Order Info
Crimper and Decapper Jaws (Part No. W225616-W225628) and sturdy stand (Part No. W225630) are sold separately. For standard voltages, see cat. no. 029110001.
Specifications
Specifications
20mm Flip-cap Aluminum Seals
Crimpenstein with 20mm Crimper and Decapper Jaw-Set; Continental Europe plug
Attaching and removing aluminum seals on crimp top/serum finish bottles and vials