missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
228.95 GBP valid until 2024-12-31
Use promo code "21976" to get your promotional price.
200.88 GBP valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Use promo code "21976" to get your promotional price.
200.88 GBP valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Click to view available options
Capacity:
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Packaging:
Convenience Pk.
Convenience pack
Valu-bulk pack
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Wide mouths provide easy access to contents when using spoons or spatulas
Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
Clear-USP Type III soda-lime glass
Specifications
Capacity | 2 oz., 60 mL |
Closure Liner | Poly/Vinyl |
Closure Material | Polypropylene |
Closure Size | 53-400 mm |
Closure Type | Screw Cap |
Diameter (Metric) | 55 mm |
Height (English) | 1.8 in. |
Height (Metric) | 48 mm |
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'