missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps

Catalog Number 12349749
35.77 GBP valid until 2024-12-31
Use promo code "23261" to get your promotional price.


Click to view available options
Capacity:
1 oz., 30mL
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Packaging:
Convenience pack
Valu-bulk pack
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for solvent, chemical and sample storage

  • Narrow mouth enables liquids to be carefully poured to prevent spillage
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

  • Available in partitioned Convenience or Valu-Bulk™ cartons

Order Info

Caps can be ordered separately.

Compliance

USP Type-III soda-lime glass

Capacity 32 oz., 1000 mL
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Size 33-400 mm
Closure Type Screw Cap
Diameter (Metric) 94 mm
Height (English) 8.18 in.
Height (Metric) 208 mm
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Clear
Diameter (English) 3.7 in.
Material Flint Glass
Packaging Valu-bulk pack
Closure Color White
Certifications/Compliance EPA, FDA
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'