Learn More
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Brand: DWK Life Sciences 224193-01
217.04 GBP valid until 2024-12-31
Use promo code "21972" to get your promotional price.
Code : 03
Additional Details : Weight : 0.52000kg
Description
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among many others
Specifications
Natural | |
1000/Cs. | |
20 mm |
Aluminum | |
Complete Tear-off Cap |