Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
£102.00 - £341.00
Product Code | Brand | Type | Color | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Color | Quantity | Price | Quantity & Availability | |||
10194172
|
DWK Life Sciences
224183-01 |
Center Disc Tear-out Cap | Natural | 1000/Cs. |
£341.00
Pack of 1000 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
10022372
|
DWK Life Sciences
224182-01 |
Center Disc Tear-out Cap | Natural | 1000/Cs. |
£102.00
Pack of 1000 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10331772
|
DWK Life Sciences
224187-01 |
Center Disc Tear-out | Natural | 1000/Cs. |
£156.00
Pack of 1000 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||
11788779
|
DWK Life Sciences
224182-07 |
Center Disc Tear-out Cap | Green | 1000/Cs. |
£134.00
Pack of 1000 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10532444
|
DWK Life Sciences
224183-07 |
Center Disc Tear-out Cap | Green | 1000/Cs. |
£156.00
Pack of 1000 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||
Description
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among others
Specifications
Aluminum | |
1000/Cs. | |
Center Disc Tear-out Cap |